1-(2-Bromoethoxy)-2,4-dichlorobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: beta-Bromo-2,4-dichlorophenetole

044074-500MG 044074-1G 044074-5G
101863-990EA 96.92 USD
101863-990 101863-992 101863-994
1-(2-Bromoethoxy)-2,4-dichlorobenzene ≥95%
1-(2-Bromoethoxy)-2,4-dichlorobenzene

Hazard code(s): Irritant

Formula: C₈H₇BrCl₂O
MW: 269.95 g/mol
Boiling Pt: 120…130 °C (0.4mm)
MDL Number: MFCD00043464
CAS Number: 6954-77-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 120-130°C/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR