2-Chloro-6-hydrazinopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: (6-Chloro-2-pyridyl)hydrazine

044032-5G 044032-25G 044032-100G
101863-774EA 15.38 USD
101863-774 102663-712 102663-714
2-Chloro-6-hydrazinopyridine ≥95%
2-Chloro-6-hydrazinopyridine

Hazard code(s): Irritant

Formula: C₅H₆ClN₃
Melting Pt: 118 °C
MDL Number: MFCD00214724
CAS Number: 5193-03-3

Order Now

Specification Test Results

Melting point 118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR