1-(3-Bromophenyl)pyrrole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(3-Bromophenyl)-1H-pyrrole, N-(3-Bromophenyl)pyrrole

043997-500MG 043997-1G 043997-5G
101863-594EA 181.54 USD
101863-594 101863-596 101863-598
1-(3-Bromophenyl)pyrrole ≥95%
1-(3-Bromophenyl)pyrrole

Hazard code(s): Irritant

Formula: C₁₀H₈BrN
MW: 222.08 g/mol
Melting Pt: 61…63 °C
MDL Number: MFCD02665243
CAS Number: 107302-22-7

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR