4'-Chlorobiphenyl-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-Chloro[1,1'-biphenyl]-4-carboxylic acid, 4'-Chloro-[1,1'-biphenyl]-4-carboxylic acid

043961-500MG 043961-1G 043961-5G
101863-424EA 747.69 USD
101863-424 101863-426 101863-428
4'-Chlorobiphenyl-4-carboxylic acid ≥95%
4'-Chlorobiphenyl-4-carboxylic acid

Hazard code(s): Irritant

Formula: C₁₃H₉ClO₂
MW: 232.67 g/mol
Melting Pt: 293…296 °C
MDL Number: MFCD03424592
CAS Number: 5748-41-4

Order Now

Specification Test Results

Melting point 293-296°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR