Methyl-4'-bromobiphenyl-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4'-bromo[1,1'-biphenyl]-4-carboxylate, Methyl 4'-bromo-[1,1'-biphenyl]-4-carboxylate
043946-1G
043946-5G
101863-340EA
430.77
USD
101863-340
101863-342
Methyl-4'-bromobiphenyl-4-carboxylate ≥95%
Methyl-4'-bromobiphenyl-4-carboxylate
Hazard code(s): Irritant
Formula:
C₁₄H₁₁BrO₂ MW: 291.14 g/mol Melting Pt: 146…148 °C |
MDL Number:
MFCD00496638 CAS Number: 89901-03-1 |
Specification Test Results
Melting point | 146-148°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...