4-Amino-5-bromo-2-methoxybenzoic acid ≥95%

Supplier: Matrix Scientific

043906-500MG 043906-1G
101863-152EA 561.54 USD
101863-152 101863-154
4-Amino-5-bromo-2-methoxybenzoic acid ≥95%
4-Amino-5-bromo-2-methoxybenzoic acid

Hazard code(s): Irritant

Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
Melting Pt: 186…188 °C
MDL Number: MFCD02186027
CAS Number: 35290-97-2
EINECS: 252-491-8

Order Now

Specification Test Results

Melting point 186-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR