ω-Bromo-2-chloroacetophenone ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: α-Bromo-2-chloroacetophenone, Bromomethyl o-chlorophenyl ketone, 2-Bromo-1-(2-chlorophenyl)ethanone, 2-Bromo-2'-chloroacetophenone, 2-Chlorophenacyl bromide

043738-1G 043738-5G 043738-25G
101862-328EA 21.54 USD
101862-328 101862-330 101862-332
ω-Bromo-2-chloroacetophenone ≥95% for synthesis
ω-Bromo-2-chloroacetophenone

Hazard code(s): Toxic, Corrosive, Lachrymator, Refrigerate

Formula: C₈H₆BrClO
Boiling Pt: 105 °C (1 mm)
Density: 1.602 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00832993
CAS Number: 5000-66-8
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR