3-Bromo-7-nitro-1H-indazole ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-7-nitro-1H-indazole, 3-Bromo-7-nitroindazole
043703-1G
043703-5G
101862-144EA
35.38
USD
101862-144
101862-146
3-Bromo-7-nitro-1H-indazole ≥95%
3-Bromo-7-nitro-1H-indazole
Hazard code(s): Irritant
Formula:
C₇H₄BrN₃O₂ MW: 242.03 g/mol Melting Pt: 182 °C |
MDL Number:
MFCD00159910 CAS Number: 74209-34-0 |
Specification Test Results
Melting point | 182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...