1-(2-Bromoethoxy)-3-nitrobenzene ≥95%

Supplier: Matrix Scientific

043693-500MG 043693-1G 043693-5G
101862-092EA 181.54 USD
101862-092 101862-094 101862-096
1-(2-Bromoethoxy)-3-nitrobenzene ≥95%
1-(2-Bromoethoxy)-3-nitrobenzene

Hazard code(s): Irritant

Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
Melting Pt: 25 °C
MDL Number: MFCD07783648
CAS Number: 13831-59-9

Order Now

Specification Test Results

Melting point 25°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR