2-Acetyl-3-chlorothiophene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(3-Chloro-2-thienyl)-1-ethanone

043479-500MG 043479-1G 043479-5G
101861-010EA 375.38 USD
101861-010 101861-012 101861-014
2-Acetyl-3-chlorothiophene ≥95%
2-Acetyl-3-chlorothiophene

Hazard code(s): Irritant

Formula: C₆H₅ClOS
MW: 160.62 g/mol
MDL Number: MFCD00794248
CAS Number: 89581-82-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR