Fenclorim ≥95%

Supplier: Matrix Scientific

Synonyms: 4,6-Dichloro-2-phenylpyrimidine

043477-500MG 043477-1G 043477-5G
101860-998EA 278.46 USD
101860-998 101861-000 101861-002
Fenclorim ≥95%
Fenclorim

Hazard code(s): Irritant

Formula: C₁₀H₆Cl₂N₂
Melting Pt: 92…93 °C
MDL Number: MFCD00754193
CAS Number: 3740-92-9

Order Now

Specification Test Results

Melting point 92-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR