2-Bromo-N-(2,6-dimethylphenyl)acetamide ≥95%

Supplier: Matrix Scientific

043445-500MG 043445-1G 043445-5G
101860-828EA 60 USD
101860-828 101860-830 101860-832
2-Bromo-N-(2,6-dimethylphenyl)acetamide ≥95%
2-Bromo-N-(2,6-dimethylphenyl)acetamide

Hazard code(s): Irritant

Formula: C₁₀H₁₂BrNO
Melting Pt: 148…151 °C
MDL Number: MFCD00215011
CAS Number: 40251-98-7

Order Now

Specification Test Results

Melting point 148-151°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR