3,4-Dichlorobenzhydrazide ≥95%
Supplier: Matrix Scientific
Synonyms:
3,4-Dichlorobenzenecarbohydrazide, 3,4-Dichlorobenzohydrazide, 3,4-Dichlorobenzene-1-carbohydrazide, 3,4-Dichlorobenzoic acid hydrazide, 3,4-Dichlorobenzoic hydrazide
043379-500MG
043379-1G
043379-5G
101860-494EA
278.46
USD
101860-494
101860-496
101860-498
3,4-Dichlorobenzhydrazide ≥95%
3,4-Dichlorobenzhydrazide
Hazard code(s): Irritant
Formula:
C₇H₆Cl₂N₂O MW: 205.04 g/mol Melting Pt: 157…160 °C |
MDL Number:
MFCD00017057 CAS Number: 28036-91-1 |
Specification Test Results
Melting point | 157-160°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...