ω-Bromo-3-cyanoacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(2-Bromoacetyl)benzonitrile, 2-Bromo-3'-cyanoacetophenone, 3-(Bromoacetyl)benzonitrile, 3-Cyanophenacyl bromide, Bromomethyl m-cyanophenyl ketone, α-Bromo-3-cyanoacetophenone, 3-(2-Bromoacetyl)benzenecarbonitrile

043362-1G 043362-5G
101860-440EA 73.85 USD
101860-440 101860-442
ω-Bromo-3-cyanoacetophenone ≥95%
ω-Bromo-3-cyanoacetophenone

Hazard code(s): Irritant

Formula: C₉H₆BrNO
MW: 224.06 g/mol
Melting Pt: 70…71 °C
MDL Number: MFCD00833267
CAS Number: 50916-55-7

Order Now

Specification Test Results

Melting point 70-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR