Methyl-2-amino-5-bromonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-5-bromopyridine-3-carboxylate

043330-1G 043330-5G
101860-276EA 15.38 USD
101860-276 101860-278
Methyl-2-amino-5-bromonicotinate ≥95%
Methyl-2-amino-5-bromonicotinate

Hazard code(s): Irritant

Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
Melting Pt: 143…146 °C
MDL Number: MFCD04117787
CAS Number: 50735-34-7

Order Now

Specification Test Results

Melting point 143-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR