1-Phenyloxindole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Phenyl-1,3-dihydro-2H-indol-2-one

043324-500MG 043324-1G 043324-5G
101860-240EA 278.46 USD
101860-240 101860-242 101860-244
1-Phenyloxindole ≥95%
1-Phenyloxindole

Hazard code(s): Irritant

Formula: C₁₄H₁₁NO
MW: 209.25 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD00234850
CAS Number: 3335-98-6

Order Now

Specification Test Results

Melting point 121-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR