6-Nitroquinoxaline ≥95%

Supplier: Matrix Scientific

043287-500MG 043287-1G 043287-5G
101860-068EA 375.38 USD
101860-068 101860-070 101860-072
6-Nitroquinoxaline ≥95%
6-Nitroquinoxaline

Hazard code(s): Irritant

Formula: C₈H₅N₃O₂
MW: 175.15 g/mol
Melting Pt: 169…171 °C
MDL Number: MFCD00462822
CAS Number: 6639-87-8

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR