4'-Cyanobiphenyl-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-Cyano[1,1'-biphenyl]-4-carboxylic acid, 4'-Cyano-[1,1'-biphenyl]-4-carboxylic acid

043263-500MG 043263-1G 043263-5G
101859-962EA 303.08 USD
101859-962 101859-964 101859-966
4'-Cyanobiphenyl-4-carboxylic acid ≥95%
4'-Cyanobiphenyl-4-carboxylic acid

Hazard code(s): Irritant

Formula: C₁₄H₉NO₂
MW: 223.23 g/mol
Melting Pt: 263…266 °C
MDL Number: MFCD00438676
CAS Number: 5728-46-1

Order Now

Specification Test Results

Melting point 263-266°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR