6-Phenoxy-3-formylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Phenoxynicotinaldehyde, 6-Phenoxypyridine-3-carboxaldehyde

042838-500MG 042838-1G 042838-5G
101857-790EA 224.62 USD
101857-790 101857-792 101857-794
6-Phenoxy-3-formylpyridine ≥95%
6-Phenoxy-3-formylpyridine

Hazard code(s): Irritant

Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD02681950
CAS Number: 173282-69-4

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR