3-Bromothiophene-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromothiophene-2-carboxaldehyde, 3-Bromo-2-formylthiophene

042834-1G 042834-25G
101857-772EA 29.23 USD
101857-772 101857-776
3-Bromothiophene-2-carbaldehyde ≥95%
3-Bromothiophene-2-carbaldehyde

Hazard code(s): Irritant

Formula: C₅H₃BrOS
MW: 191.05 g/mol
Boiling Pt: 91 °C (3mm)
MDL Number: MFCD00126680
CAS Number: 930-96-1

Order Now

Specification Test Results

Boiling point 91°C/3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR