ω-Bromo-3-chloroacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3'-chloroacetophenone, Bromomethyl m-chlorophenyl ketone, 2-Bromo-1-(3-chlorophenyl)ethanone, 3-Chlorophenacyl bromide, α-Bromo-3-chloroacetophenone

042825-5G 042825-25G
101857-746EA 107.69 USD
101857-746 102833-118
ω-Bromo-3-chloroacetophenone ≥95%
ω-Bromo-3-chloroacetophenone

Hazard code(s): Irritant

Formula: C₈H₆BrClO
MW: 233.49 g/mol
Melting Pt: 40…42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792669
CAS Number: 41011-01-2

Order Now

Specification Test Results

Melting point 40-42°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR