2-Acetyl-4-bromothiophene ≥95%
Supplier: Matrix Scientific
042795-1G
042795-5G
101857-568EA
36.92
USD
101857-568
101857-570
2-Acetyl-4-bromothiophene ≥95%
2-Acetyl-4-bromothiophene
Hazard code(s): Irritant
Formula:
C₆H₅BrOS MW: 205.08 g/mol Boiling Pt: 94 °C (1.8mm) |
MDL Number:
MFCD01566552 CAS Number: 7209-11-2 |
Specification Test Results
Boiling point | 94°C/1.8mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...