Methyl-5-bromo-2-hydroxynicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-2-oxo-1,2-dihydropyridine-3-carboxylate, methyl 5-bromo-2-hydroxypyridine-3-carboxylate, Methyl 5-bromo-2-oxo-1,2-dihydro-3-pyridinecarboxylate

042778-1G 042778-5G
101857-484EA 38.46 USD
101857-484 101857-486
Methyl-5-bromo-2-hydroxynicotinate ≥95%
Methyl-5-bromo-2-hydroxynicotinate

Hazard code(s): Irritant

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Melting Pt: 181…183 °C
MDL Number: MFCD03844843
CAS Number: 120034-05-1

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR