3-Amino-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 1H-Indazol-3-amine, Indazol-3-amine, 1H-Indazol-3-ylamine, Indazol-3-ylamine, 3-Amino-1H-indazole

042721-1G 042721-5G
101857-184EA 23.08 USD
101857-184 101857-186
3-Amino-1H-indazole ≥95%
3-Amino-1H-indazole

Hazard code(s): Irritant

Formula: C₇H₇N₃
MW: 133.15 g/mol
Melting Pt: 156…157 °C
MDL Number: MFCD00182045
CAS Number: 874-05-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 156-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR