6,7-Dimethoxy-4-quinazolone ≥95%
Supplier: Matrix Scientific
Synonyms:
6,7-Dimethoxy-4(3H)-quinazolinone, 6,7-Dimethoxy-1H-quinazolin-4-one
042714-1G
042714-5G
042714-25G
101857-148EA
15.38
USD
101857-148
101857-150
101857-152
6,7-Dimethoxy-4-quinazolone ≥95%
6,7-Dimethoxy-4-quinazolone
Hazard code(s): Irritant
Formula:
C₁₀H₁₀N₂O₃ MW: 206.2 g/mol Melting Pt: 296…297 °C |
MDL Number:
MFCD01570147 CAS Number: 13794-72-4 |
Specification Test Results
Melting point | 296-297°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...