4-Chlorobenzaldehyde oxime ≥95%
Supplier: Matrix Scientific
042690-1G
042690-5G
101857-026EA
67.69
USD
101857-026
101857-028
4-Chlorobenzaldehyde oxime ≥95%
4-Chlorobenzaldehyde oxime
Hazard code(s): Irritant
Formula:
C₇H₆ClNO Melting Pt: 108…110 °C |
MDL Number:
MFCD00016412 CAS Number: 3848-36-0 |
Specification Test Results
Melting point | 108-110°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...