Methyl-4-amino-2-methoxybenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-amino-2-methoxybenzenecarboxylate, 4-Amino-2-methoxybenzoic acid methyl ester, Methyl 4-amino-2-methoxybenzoate
042643-25G
042643-100G
101856-770EA
33.85
USD
101856-770
101856-772
Methyl-4-amino-2-methoxybenzoate ≥95%
Methyl-4-amino-2-methoxybenzoate
Hazard code(s): Irritant
Formula:
C₉H₁₁NO₃ MW: 181.19 g/mol Melting Pt: 147…150 °C |
MDL Number:
MFCD00017202 CAS Number: 27492-84-8 EINECS: 248-494-9 |
Specification Test Results
Melting point | 147-150°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...