3-(Trifluoromethyl)benzamidoxime ≥95%

Supplier: Matrix Scientific

Synonyms: N'-Hydroxy-3-(trifluoromethyl)-benzenecarboximidamide, N-Hydroxy-3-(trifluoromethyl)benzimidamide

042610-500MG 042610-1G 042610-5G
101856-598EA 278.46 USD
101856-598 101856-600 101856-602
3-(Trifluoromethyl)benzamidoxime ≥95%
3-(Trifluoromethyl)benzamidoxime

Hazard code(s): Irritant

Formula: C₈H₇F₃N₂O
MW: 204.15 g/mol
Melting Pt: 87…89 °C
MDL Number: MFCD02647312
CAS Number: 40067-80-9

Order Now

Specification Test Results

Melting point 87-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR