Methyl-3,4-dichlorobenzoate ≥95%

Supplier: Matrix Scientific

042596-500MG 042596-1G 042596-5G
101856-530EA 181.54 USD
101856-530 101856-532 101856-534
Methyl-3,4-dichlorobenzoate ≥95%
Methyl-3,4-dichlorobenzoate

Hazard code(s): Irritant

Formula: C₈H₆Cl₂O₂
Melting Pt: 46…48 °C
MDL Number: MFCD00018165
CAS Number: 2905-68-2

Order Now

Specification Test Results

Melting point 46-48°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR