1-(4-Aminophenyl)-4-methylpiperazine ≥97%
Supplier: Matrix Scientific
Synonyms:
4-(4-Methylpiperazino)aniline, 4-(4-Methylpiperazin-1-yl)phenylamine, 4-(4-Methyl-1-piperazinyl)aniline
042562-1G
042562-5G
101856-372EA
15.38
USD
101856-372
101856-374
1-(4-Aminophenyl)-4-methylpiperazine ≥97%
1-(4-Aminophenyl)-4-methylpiperazine
Hazard code(s): Irritant
Formula:
C₁₁H₁₇N₃ MW: 191.28 g/mol Melting Pt: 95…97 °C |
MDL Number:
MFCD00172703 CAS Number: 16153-81-4 UN: 3263 ADR: 8,III |
Specification Test Results
Melting point | 95-97°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...