3-Phenylpyridine ≥98%

Supplier: Matrix Scientific
Warning

042492-5G 042492-25G 042492-100G
101856-014EA 33.85 USD
101856-014 101856-016 101856-018
3-Phenylpyridine ≥98%
3-Phenylpyridine

Hazard code(s): Irritant

Formula: C₁₁H₉N
Boiling Pt: 273 - 274 °C (1013 hPa)
Melting Pt: 76 - 77 °C
Density: 1.08 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006380
CAS Number: 1008-88-4
EINECS: 213-762-6

Order Now

Specification Test Results

Boiling point 272°C
Density 1.085
ND20 1.616


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR