L(+)-Asparagine, anhydrous ≥95%

Supplier: Matrix Scientific

041292-25G 041292-100G
101851-232EA 40 USD
101851-232 101851-234
L(+)-Asparagine, anhydrous ≥95%
L(+)-Asparagine

Hazard code(s): Irritant

Formula: H₂NCOCH₂CH(NH₂)CO₂H
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 234…236 °C
Storage Temperature: Ambient
MDL Number: MFCD00064401
CAS Number: 70-47-3
EINECS: 200-735-9
Merck Index: 12,00872
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=3d09743d0834d64e4719e8295f634c39195c571f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR