2-Fluoro-3-hydroxypyridine ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Fluoropyridin-3-ol, 2-Fluoro-3-pyridinol

041254-5G 041254-25G
101851-062EA 53.85 USD
101851-062 101851-064
2-Fluoro-3-hydroxypyridine ≥98% for synthesis
2-Fluoro-3-hydroxypyridine

Hazard code(s): Irritant

Formula: C₅H₄FNO
MDL Number: MFCD04112569
CAS Number: 174669-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR