2,4,5-Tribromotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-2,4,5-tribromobenzene, 1,2,4-Tribromo-5-methylbenzene

041248-1G 041248-5G 041248-25G
101851-038EA 21.54 USD
101851-038 101851-040 101851-042
2,4,5-Tribromotoluene ≥98%
2,4,5-Tribromotoluene

Hazard code(s): Irritant

Formula: C₇H₅Br₃
MW: 328.83 g/mol
MDL Number: MFCD00052984
CAS Number: 3278-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR