1-Boc-3-hydroxypiperidine ≥97%

Supplier: Matrix Scientific

Synonyms: tert-butyl 3-hydroxypiperidine-1-carboxylate

041245-5G 041245-25G
101851-028EA 15.38 USD
101851-028 101851-030
1-Boc-3-hydroxypiperidine ≥97%
1-Boc-3-hydroxypiperidine

Hazard code(s): Irritant

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 66…70 °C
MDL Number: MFCD02093938
CAS Number: 85275-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR