4-Bromosalicylaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-hydroxybenzaldehyde

041221-1G 041221-5G
101850-962EA 27.69 USD
101850-962 101850-964
4-Bromosalicylaldehyde ≥98%
4-Bromosalicylaldehyde

Hazard code(s): Irritant

Formula: C₇H₅BrO₂
MW: 201.02
Melting Pt: 50…54 °C
MDL Number: MFCD06252606
CAS Number: 22532-62-3

Order Now

Specification Test Results

Melting point 50-54°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR