3,5-Dibromotoluene ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 1,3-Dibromo-5-methylbenzene

041118-25G 041118-100G
101850-840EA 44.62 USD
101850-840 101850-842
3,5-Dibromotoluene ≥97%
3,5-Dibromotoluene

Hazard code(s): Irritant

Formula: C₇H₆Br₂
MW: 249.93 g/mol
Melting Pt: 37…40 °C
MDL Number: MFCD00013528
CAS Number: 1611-92-3

Order Now

Specification Test Results

Melting point 37-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR