2-Fluoro-4-iodopyridine ≥98%

Supplier: Matrix Scientific
Warning

041117-1G 041117-5G 041117-25G
101850-834EA 15.38 USD
101850-834 101850-836 101850-838
2-Fluoro-4-iodopyridine ≥98%
2-Fluoro-4-iodopyridine

Hazard code(s): Irritant

Formula: C₅H₃FIN
MW: 222.99 g/mol
Melting Pt: 57…58 °C
MDL Number: MFCD03092926
CAS Number: 22282-70-8

Order Now

Specification Test Results

Melting point 57-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR