2-Fluoro-6-methoxypyridine ≥98%

Supplier: Matrix Scientific

041031-1G 041031-5G
101850-656EA 44.62 USD
101850-656 101850-658
2-Fluoro-6-methoxypyridine ≥98%
2-Fluoro-6-methoxypyridine

Hazard code(s): Irritant

Formula: C₆H₆FNO
MDL Number: MFCD09998161
CAS Number: 116241-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR