2-Methylnicotinic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Methylpyridine-3-carboxylic acid, 2-Methyl-3-pyridinecarboxylic acid
041028-1G
041028-5G
101850-648EA
21.54
USD
101850-648
101850-650
2-Methylnicotinic acid ≥98%
2-Methylnicotinic acid
Hazard code(s): Irritant
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Melting Pt: 228…230 °C |
MDL Number:
MFCD00013449 CAS Number: 3222-56-8 |
Specification Test Results
Melting point | 228-230°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...