3-Methylpicolinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Methylpyridine-2-carboxylic acid

041026-5G 041026-25G
101850-644EA 36.92 USD
101850-644 101850-646
3-Methylpicolinic acid ≥98%
3-Methylpicolinic acid

Hazard code(s): Irritant

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 114…118 °C
MDL Number: MFCD00191553
CAS Number: 4021-07-2

Order Now

Specification Test Results

Melting point 114-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR