3-Fluoro-2-methoxypyridine ≥98% for synthesis

Supplier: Matrix Scientific

041025-1G 041025-5G 041025-25G
101850-638EA 53.85 USD
101850-638 101850-640 101850-642
3-Fluoro-2-methoxypyridine ≥98% for synthesis
3-Fluoro-2-methoxypyridine

Hazard code(s): Irritant

Formula: C₆H₆FNO
MDL Number: MFCD03094945
CAS Number: 884494-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR