3-Chloro-6-methoxypyridazine
Supplier: Matrix Scientific
|
Warning
|
040927-5G
040927-25G
101850-438EA
267.69
USD
101850-438
101850-440
3-Chloro-6-methoxypyridazine
3-Chloro-6-methoxypyridazine
Formula:
C₅H₅ClN₂O Boiling Pt: 174…175 °C (3 mmHg) Melting Pt: 91 °C |
MDL Number:
MFCD00006467 CAS Number: 1722-10-7 EINECS: 217-019-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...