Sulfamide ≥97%

Supplier: Matrix Scientific

039968-25G 039968-100G
101848-572EA 35.38 USD
101848-572 101848-574
Sulfamide ≥97%
Sulphamide

Hazard code(s): Irritant

Formula: H₄N₂O₂S
MDL Number: MFCD00011606
CAS Number: 7803-58-9
EINECS: 232-262-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR