Methyl propiolate ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester

039967-25G 039967-100G
101848-568EA 73.85 USD
101848-568 101848-570
Methyl propiolate ≥97% for synthesis
Methyl propiolate

Hazard code(s): Flammable, Irritant

Formula: C₄H₄O₂
Boiling Pt: 105 °C (1013 hPa)
Density: 1.196 g/cm³ (20 °C)
MDL Number: MFCD00008572
CAS Number: 922-67-8
EINECS: 213-083-5
UN: 3272
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR