Tetrahydro-4H-pyran-4-one ≥95%

Supplier: Matrix Scientific

Synonyms: Dihydro-2H-pyran-4(3H)-one, Tetrahydro-α-pyran-4-one, Tetrahydro-2H-pyran-4-one, Tetrahydro-γ-pyran-4-one

039953-25G 039953-100G
101848-532EA 32.31 USD
101848-532 101848-534
Tetrahydro-4H-pyran-4-one ≥95%
Tetrahydro-4H-pyran-4-one

Hazard code(s): Irritant

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 165…167 °C
Density: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00006581
CAS Number: 29943-42-8
EINECS: 249-967-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR