4-Chloroisatoic anhydride

Supplier: Matrix Scientific

Synonyms: 7-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione, 7-Chloro-2H-3,1-benzoxazine-2,4(1H)-dione

039859-5G 039859-25G
101848-336EA 89.23 USD
101848-336 101848-338
4-Chloroisatoic anhydride
4-Chloroisatoic anhydride
Formula: C₈H₄ClNO₃
MW: 197.58 g/mol
MDL Number: MFCD00090431
CAS Number: 40928-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR