4-(Trifluoromethyl)phenyl isocyanate ≥95%

Supplier: Matrix Scientific
Danger

Synonyms: α,α,α-Trifluoro-p-tolyl isocyanate, 1-Isocyanato-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)phenylisocyanate, p-Trifluoromethylphenylisocyanate

038715-25G 038715-100G
101846-064EA 230.77 USD
101846-064 101846-066
4-(Trifluoromethyl)phenyl isocyanate ≥95%
4-(Trifluoromethyl)phenyl isocyanate
Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Boiling Pt: 205 - 207 °C (1013 hPa)
Melting Pt: 29 - 31 °C
MDL Number: MFCD00002028
CAS Number: 1548-13-6
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR