4-Methoxyphenyl isocyanate

Supplier: Matrix Scientific

Synonyms: 1-Isocyanato-4-methoxybenzene

038314-25G 038314-100G
101845-252EA 355.38 USD
101845-252 101845-254
4-Methoxyphenyl isocyanate
4-Methoxyphenyl isocyanate

Hazard code(s): Irritant

Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 106…110 °C
Density: 1.1559 g/cm³ (20 °C)
MDL Number: MFCD00002026
CAS Number: 5416-93-3
EINECS: 226-513-1
UN: 2206
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 106-110 ºC
Density 1.151

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR