4-Hydroxy-2-methoxybenzaldehyde ≥95%

Supplier: Matrix Scientific
Warning

037590-1G 037590-25G
101843-726EA 49.23 USD
101843-726 101843-730
4-Hydroxy-2-methoxybenzaldehyde ≥95%
4-Hydroxy-2-methoxybenzaldehyde

Hazard code(s): Irritant

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00051964
CAS Number: 18278-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR